Assay Details
Gene Symbol: Ddx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025860.3 | NP_080136.2 | 3 - 4 | 588 | 88 | |
GenBank mRNA | AK019845.1 | - | 3 - 4 | 512 | 88 | |
AK075864.1 | - | 3 - 4 | 525 | 88 | ||
BC017160.1 | - | 3 - 4 | 510 | 88 | ||
BC028246.1 | - | 3 - 4 | 559 | 2582151 | 88 | |
BC103776.1 | - | 3 - 4 | 525 | 6492142 | 88 | |
BC141230.1 | - | 3 - 4 | 527 | 9055842 | 88 | |
BC141231.1 | - | 3 - 4 | 527 | 9055843 | 88 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
