Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100023965.mm.1 |
NM_025860.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023965.mm.1 |
NM_025860.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023965.mm.1 |
TC0100002974.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100023963.mm.1 |
NM_025860.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023963.mm.1 |
NM_025860.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023963.mm.1 |
TC0100002974.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100023964.mm.1 |
NM_025860.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023964.mm.1 |
NM_025860.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100023964.mm.1 |
TC0100002974.mm.1 |
MTA TC |