Assay Details
Gene Symbol: Ddx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025860.3 | NP_080136.2 | 9 - 10 | 1592 | 103 | |
GenBank mRNA | AK019845.1 | - | 9 - 10 | 1516 | 103 | |
AK210635.1 | - | 1 - 2 | 97 | 103 | ||
BC017160.1 | - | 9 - 10 | 1512 | 103 | ||
BC028246.1 | - | 9 - 10 | 1563 | 2582151 | 103 | |
BC103776.1 | - | 9 - 10 | 1529 | 6492142 | 103 | |
BC141230.1 | - | 9 - 10 | 1531 | 9055842 | 103 | |
BC141231.1 | - | 9 - 10 | 1531 | 9055843 | 103 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
