Assay Details
Gene Symbol: Mrpl48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198831.2 | NP_942128.2 | 3 - 4 | 235 | 84 | |
NR_003559.1 | 3 - 4 | 200 | 84 | |||
GenBank mRNA | AK007653.1 | - | 3 - 4 | 506 | 84 | |
AK019161.1 | - | 3 - 4 | 197 | 84 | ||
AK171917.1 | - | 3 - 4 | 208 | 84 | ||
AK211110.1 | - | 2 - 3 | 97 | 84 | ||
BC037190.1 | - | 3 - 4 | 190 | 4168034 | 84 | |
BC116358.1 | - | 2 - 3 | 105 | 40099962 | 84 | |
BC116359.1 | - | 2 - 3 | 102 | 40099963 | 84 | |
BC128487.1 | - | 2 - 3 | 105 | 40099964 | 84 | |
BC128488.1 | - | 2 - 3 | 105 | 40099965 | 84 | |
BG077486.2 | - | 2 - 3 | 69 | 84 | ||
BU604260.1 | - | 3 - 4 | 228 | 84 | ||
CD363582.1 | - | 3 - 4 | 166 | 84 | ||
DN174444.1 | - | 3 - 4 | 235 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC116358.1,BC116359.1,BC128487.1,BC128488.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
