Assay Details
Gene Symbol: Mrpl48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198831.2 | NP_942128.2 | 6 - 7 | 592 | 102 | |
NR_003559.1 | 6 - 7 | 557 | 102 | |||
GenBank mRNA | AK007653.1 | - | 6 - 7 | 863 | 101 | |
AU044645.1 | - | 2 - 3 | 102 | |||
BU604260.1 | - | 6 - 7 | 585 | 102 | ||
CD363582.1 | - | 6 - 7 | 523 | 102 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
