Assay Details
| Gene Symbol: Olfr73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054090.1 | NP_473431.1 | 1 | 640 | 120 | |
| GenBank mRNA | AB061228.1 | - | 1 | 640 | 120 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P316199 | NM_054090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P316199 | AB061228.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P316199 | NM_054090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P316199 | AB061228.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154571 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P316199 | NM_054090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P316199 | AB061228.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P316199 | NM_054090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P316199 | AB061228.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P316199 | NM_054090.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P316199 | AB061228.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P316199 | NM_054090.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P316199 | AB061228.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154571 | - | - |
| Whole Mouse Genome G4122A | A_51_P316199 | NM_054090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316199 | AB061228.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316199 | NM_054090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316199 | AB061228.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130482 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02036267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02036266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02036275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01490648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21408292 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484711 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421777_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484710 | - | - |
| Mouse Genome 430 2.0 Array | 1421777_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421777_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 117004_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 117004_at | - | - |
| Mouse Expression Set 430 | 1421777_at | - | - |
| Mouse Expression Set 430 | 1421777_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421777_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421777_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421777_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421777_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 859313 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3850278 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3850278 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003850278 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2668756 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2668756 | - | - |
| mouse-6 v1.1 (Illumina) | scl19038.1.1_199-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3850278 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl19038.1.1_199-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850278 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2668756 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2668756 | - | - |

Olfr73
Genomic Map