Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200033861.mm.1 |
NM_178250.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033861.mm.1 |
NM_178250.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033861.mm.1 |
TC0200003980.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200033862.mm.1 |
NM_178250.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033862.mm.1 |
NM_178250.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033862.mm.1 |
TC0200003980.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200033864.mm.1 |
NM_178250.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033864.mm.1 |
NM_178250.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033864.mm.1 |
TC0200003980.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200033863.mm.1 |
NM_178250.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033863.mm.1 |
NM_178250.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033863.mm.1 |
TC0200003980.mm.1 |
MTA TC |