Assay Details
| Gene Symbol: Ulk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013881.4 | NP_038909.3 | 17 - 18 | 2039 | 72 | |
| GenBank mRNA | AB019577.1 | - | 17 - 18 | 1564 | 72 | |
| AF145922.1 | - | 17 - 18 | 1932 | 72 | ||
| AK122331.2 | - | 17 - 18 | 2021 | 72 | ||
| AK146620.1 | - | 17 - 18 | 2001 | 72 | ||
| AK146893.1 | - | 17 - 18 | 2040 | 72 | ||
| AK148142.1 | - | 17 - 18 | 2040 | 72 | ||
| BC013691.1 | - | 15 - 16 | 1289 | 72 | ||
| BC046778.1 | - | 17 - 18 | 1997 | 5709559 | 72 | |
| BC053029.1 | - | 17 - 18 | 1997 | 5709559 | 72 | |
More Information
Set Membership: |
|

Ulk2
Genomic Map