Assay Details
| Gene Symbol: Drg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021354.1 | NP_067329.1 | 9 - 10 | 870 | 67 | |
| GenBank mRNA | AJ243590.1 | - | 9 - 10 | 870 | 67 | |
| AK012988.1 | - | 9 - 10 | 838 | 67 | ||
| BC082564.1 | - | 9 - 10 | 826 | 30551418 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Drg2
Genomic Map