Assay Details
| Gene Symbol: Mrpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026490.2 | NP_080766.1 | 3 - 4 | 495 | 106 | |
| GenBank mRNA | AK021180.1 | - | 3 - 4 | 489 | 106 | |
| AK030793.1 | - | 3 - 4 | 483 | 106 | ||
| AK050637.1 | - | 3 - 4 | 494 | 106 | ||
| AK141554.1 | - | 3 - 4 | 505 | 106 | ||
| BC020315.2 | - | 3 - 4 | 471 | 106 | ||
| BC043921.1 | - | 3 - 4 | 476 | 5361515 | 106 | |
More Information
Set Membership: |
|

Mrpl19
Genomic Map