Assay Details
Gene Symbol: Gpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290742.1 | NP_001277671.1 | 3 - 4 | 743 | 114 | |
NM_133884.2 | NP_598645.2 | 3 - 4 | 812 | 114 | ||
GenBank mRNA | BC018407.1 | - | 3 - 4 | 805 | 4015340 | 114 |
CN721622.1 | - | 3 - 4 | 414 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23318322 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23318323 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23318321 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23318324 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23318326 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055291 | BC018407.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055291 | BQ552936.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055291 | NM_133884.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015430 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184106 | BC018407.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184106 | BQ552936.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184106 | CN721622.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184106 | NM_133884.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143659 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527541 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483693 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 37755 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728862 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 245876 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139274 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184106 | BC018407.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184106 | BQ552936.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184106 | CN721622.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184106 | NM_133884.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055291 | BC018407.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055291 | BQ552936.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2055291 | NM_133884.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015430 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23318324 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23318327 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23318335 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23318334 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23318323 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23318333 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23318338 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23318329 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23318332 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23318331 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23318321 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00993374 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139274 | - | - |
Whole Mouse Genome G4122A | A_51_P241111 | BC018407.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101259 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113802 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139274 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03865149 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03865143 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23318324 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23318322 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23318321 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23318326 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23318323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23318361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318324 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03543365 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318349 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03895221 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04096787 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03895215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318380 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03603727 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318348 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03895219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03895222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03543361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03543363 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03895220 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03895226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318352 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03740016 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03895214 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03978819 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318358 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318360 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318388 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03740009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318342 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318382 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318343 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03838963 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318386 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03832660 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318359 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318378 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318321 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04084818 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318334 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318327 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03740007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04093261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318347 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318389 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03919340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03895218 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23318356 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03665876 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10508906 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10508903 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10508902 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10508904 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10508905 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10508901 | - | - |
Mouse Genome 430 2.0 Array | 1448836_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1448836_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100210_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100210_at | - | - |
Mouse Expression Set 430 | 1448836_s_at | - | - |
Mouse Expression Set 430 | 1448836_PM_s_at | - | - |
Murine Genome U74v2 Set | 104473_PM_at | - | - |
Murine Genome U74v2 Set | 104473_at | - | - |
Mouse Genome 430A 2.0 Array | 1448836_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1448836_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6980537 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6980537 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006980537 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214375 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1214375 | - | - |
mouse-6 v1.1 (Illumina) | scl24858.5.1_216-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6980537 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl24858.5.1_216-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980537 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1214375 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214375 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl24858.5.1_216-S | - | - |
