Assay Details
| Gene Symbol: Prokr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144944.3 | NP_659193.3 | 2 - 3 | 769 | 96 | |
| GenBank mRNA | AF487279.1 | - | 1 - 2 | 447 | 96 | |
| AK041586.1 | - | 2 - 3 | 693 | 96 | ||
| AK080980.1 | - | 2 - 3 | 720 | 96 | ||
| BC043116.1 | - | 2 - 3 | 722 | 6408655 | 96 | |
| BC057557.1 | - | 2 - 3 | 722 | 6408655 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prokr2
Genomic Map