Assay Details
Gene Symbol: Olfr986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146615.2 | NP_666826.1 | 1 | 889 | 111 | |
GenBank mRNA | BC116768.1 | - | 1 | 889 | 40126087 | 111 |
BC116770.1 | - | 1 | 889 | 40126089 | 111 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094337 | BC116770.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094337 | NM_146615.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094337 | BC116768.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168217 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094337 | BC116770.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094337 | NM_146615.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094337 | BC116768.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332321 | BC116770.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332321 | NM_146615.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332321 | BC116768.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441662 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441673 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441669 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441664 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441657 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441656 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441693 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441684 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441659 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441666 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441671 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441690 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441686 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441665 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441661 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441654 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441689 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441694 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441696 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26441695 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168217 | - | - |
Whole Mouse Genome G4122A | A_51_P332321 | BC116770.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P332321 | NM_146615.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P332321 | BC116768.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115774 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168217 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07659447 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07659452 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07659456 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07627107 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07592762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26441712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441657 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07483050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441648 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07659443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441665 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441650 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07481318 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07659439 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07659447 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07611701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441661 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441638 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441681 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07611703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441643 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441669 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441652 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26441693 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10584539 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10584540 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10584538 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10584537 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6350333 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6350333 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006350333 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228050 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1228050 | - | - |
mouse-6 v1.1 (Illumina) | scl37071.3.154_246-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6350333 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl37071.3.154_246-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350333 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2891121 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2891121 | - | - |
