Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200025757.mm.1 |
NM_146116.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025757.mm.1 |
NM_146116.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025757.mm.1 |
TC0200003135.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025759.mm.1 |
NM_146116.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025759.mm.1 |
NM_146116.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025759.mm.1 |
TC0200003135.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025764.mm.1 |
NM_146116.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025764.mm.1 |
NM_146116.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025764.mm.1 |
TC0200003135.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025755.mm.1 |
NM_146116.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025755.mm.1 |
NM_146116.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025755.mm.1 |
TC0200003135.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025760.mm.1 |
NM_146116.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025760.mm.1 |
NM_146116.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025760.mm.1 |
TC0200003135.mm.1 |
MTA TC |