Assay Details
| Gene Symbol: Uqcrq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025352.2 | NP_079628.1 | 1 | 170 | 59 | |
| GenBank mRNA | AA510422.1 | - | 1 | 58 | ||
| AK002389.1 | - | 2 | 66 | 59 | ||
| AK003160.1 | - | 2 | 82 | 59 | ||
| AK005374.1 | - | 152 | 59 | |||
| AK007982.1 | - | 1 | 131 | 59 | ||
| BC027809.1 | - | 1 | 129 | 59 | ||
| BC028519.1 | - | 1 | 154 | 1365708 | 59 | |
| CB950804.1 | - | 1 | 173 | 59 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Uqcrq
Genomic Map