Assay Details
Gene Symbol: Mesdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023403.3 | NP_075892.3 | 3 | 4434 | 80 | |
GenBank mRNA | AK031949.1 | - | 3 | 4177 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25285353 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25285354 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P222522 | BC014742.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P222522 | NM_023403.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P222522 | AK031949.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P222522 | AK076760.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P222522 | BC014742.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P222522 | NM_023403.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P222522 | AK031949.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P222522 | AK076760.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 582186 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338163 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208483 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754454 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88790 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126862 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155311 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P222522 | BC014742.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P222522 | NM_023403.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P222522 | AK031949.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P222522 | AK076760.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P222522 | BC014742.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P222522 | NM_023403.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P222522 | AK031949.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P222522 | AK076760.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25285359 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25285364 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25285362 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25285354 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25285356 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25285353 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25285369 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25285368 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25285358 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25285360 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06264793 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155311 | - | - |
Whole Mouse Genome G4122A | A_51_P222522 | BC014742.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P222522 | NM_023403.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P222522 | AK031949.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P222522 | AK076760.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P222522 | BC014742.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P222522 | NM_023403.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P222522 | AK031949.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P222522 | AK076760.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P574974 | AK085084.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P574974 | NM_023403.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P574974 | AK031949.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P446363 | BC014742.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P446363 | NM_023403.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P446363 | AK031949.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P446363 | AK076760.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P446363 | AK076773.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110307 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155311 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06056863 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25285353 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25285354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25285347 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05980393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05980395 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285384 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285341 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285379 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06056863 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285335 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06012912 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06012899 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285347 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285333 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285337 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285373 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285351 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285368 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06012918 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06368125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285395 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285340 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285358 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285360 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285344 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285330 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285385 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285329 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285381 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285327 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285400 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285354 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285406 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285389 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285346 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285332 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285362 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285396 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06263305 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25285350 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10554705 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10554706 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10554707 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10554708 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10554709 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416181_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1444035_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436749_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10554704 | - | - |
Mouse Genome 430 2.0 Array | 1444035_at | - | - |
Mouse Genome 430 2.0 Array | 1444035_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1416181_at | - | - |
Mouse Genome 430 2.0 Array | 1416181_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1436749_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1436749_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67943_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67943_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700010660.mm.1 | TC0700001203.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700010660.mm.1 | NM_023403.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700010660.mm.1 | NM_023403.3 | Refseq |
Mouse Expression Set 430 | 1416181_PM_at | - | - |
Mouse Expression Set 430 | 1416181_at | - | - |
Mouse Expression Set 430 | 1444035_at | - | - |
Mouse Expression Set 430 | 1436749_at | - | - |
Mouse Expression Set 430 | 1444035_PM_at | - | - |
Mouse Expression Set 430 | 1436749_PM_at | - | - |
Murine Genome U74v2 Set | 162231_r_at | - | - |
Murine Genome U74v2 Set | 95405_PM_at | - | - |
Murine Genome U74v2 Set | 162231_PM_r_at | - | - |
Murine Genome U74v2 Set | 95405_at | - | - |
Mouse Genome 430A 2.0 Array | 1416181_at | - | - |
Mouse Genome 430A 2.0 Array | 1416181_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416181_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416181_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436749_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444035_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444035_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436749_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 393216 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 510520 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 510520 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000510520 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2709604 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2709604 | - | - |
mouse-6 v1.1 (Illumina) | scl32441.4_1-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 510520 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl32441.4_1-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510520 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2709604 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2709604 | - | - |
