Assay Details
| Gene Symbol: Hist2h3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178216.3 | NP_835734.2 | 1 | 226 | 129 | |
| GenBank mRNA | BC094041.1 | - | 226 | 5099464 | 129 | |
| Gene Symbol: Hist2h3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054045.4 | NP_473386.1 | 1 | 211 | 129 | |
| GenBank mRNA | BC015270.1 | - | 1 | 211 | 4240748 | 129 | 
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	This assay is in the 5'UTR region of the BC015270.1,NM_054045.4,NM_178216.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
| Set Membership: |  Ambion siRNA available  Amplicon length greater than or equal to 101  3' Most  Mammalian Gene Collection  Detect Genomic DNA  Mus musculus  1700  Both primers and probe map within a single exon | 
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
 Back To Top
 Back To Top
 Hist2h3c1
Hist2h3c1 Genomic Map
Genomic Map Back To Top
 Back To Top