Assay Details
| Gene Symbol: Olfr285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011778.1 | NP_001011778.1 | 1 | 915 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001305 | NM_001011778.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P125405 | NM_001011778.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931983 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001305 | NM_001011778.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P125405 | NM_001011778.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P150337 | NM_207554.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P150337 | NM_001011778.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30474080 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08476012 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08476012 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12360240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30474111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12360247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11953775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11926537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11926542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12286817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12360236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12360232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11926534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12086289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30474118 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10432151 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10432150 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 685739 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2776153 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2776153 | - | - |

Olfr285
Genomic Map