Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200045519.mm.1 |
NM_001199308.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045519.mm.1 |
TC0200005342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200045522.mm.1 |
NM_001199308.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045522.mm.1 |
TC0200005342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200045517.mm.1 |
NM_001199308.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045517.mm.1 |
TC0200005342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200045518.mm.1 |
NM_001199308.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045518.mm.1 |
TC0200005342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200045524.mm.1 |
NM_001199308.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045524.mm.1 |
TC0200005342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200045520.mm.1 |
NM_001199308.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045520.mm.1 |
TC0200005342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200045521.mm.1 |
NM_001199308.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045521.mm.1 |
TC0200005342.mm.1 |
MTA TC |