Assay Details
Gene Symbol: BC094916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024721.2 | NP_001019892.2 | 4 - 5 | 946 | 96 | |
GenBank mRNA | AK156095.1 | - | 4 - 5 | 942 | 96 | |
AK171847.1 | - | 4 - 5 | 937 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
double-stranded DNA binding
transcription factor binding
identical protein binding
poly(A) RNA binding
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
double-stranded DNA binding
transcription factor binding
identical protein binding
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082319 | NM_001024721.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709670 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403030 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96906 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082319 | NM_001024721.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20838618 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20838643 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20838620 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20838637 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20838636 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00254119 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04359097 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00254119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20838619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20838537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20838636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20838552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20838530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20838550 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838636 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838628 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838587 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838547 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838613 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838550 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838545 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838555 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838552 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838643 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838609 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20838639 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00905685 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10360371 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360372 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10360370 | - | - |
Affymetrix Mouse Clariom S Array | PSR0100029454.mm.1 | NM_001024721.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029454.mm.1 | NM_001024721.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029454.mm.1 | TC0100003590.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100029458.mm.1 | NM_001024721.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029458.mm.1 | NM_001024721.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029458.mm.1 | TC0100003590.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100029460.mm.1 | NM_001024721.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029460.mm.1 | NM_001024721.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029460.mm.1 | TC0100003590.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100029462.mm.1 | NM_001024721.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029462.mm.1 | NM_001024721.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029462.mm.1 | TC0100003590.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100029463.mm.1 | NM_001024721.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029463.mm.1 | NM_001024721.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029463.mm.1 | TC0100003590.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100029459.mm.1 | NM_001024721.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029459.mm.1 | NM_001024721.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029459.mm.1 | TC0100003590.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100029461.mm.1 | NM_001024721.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029461.mm.1 | NM_001024721.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100029461.mm.1 | TC0100003590.mm.1 | MTA TC |
Mouse Expression Set 430 | 1435330_PM_at | - | - |
Mouse Expression Set 430 | 1435330_at | - | - |
Murine Genome U74v2 Set | 103615_at | - | - |
Murine Genome U74v2 Set | 103615_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1435330_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1435330_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250523 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1250523 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3151243 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3072407 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3151243 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3072407 | - | - |
