Assay Details
| Gene Symbol: Cyp26c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105201.1 | NP_001098671.1 | 4 - 5 | 861 | 78 | |
| GenBank mRNA | BC151098.1 | - | 4 - 5 | 960 | 9088011 | 78 |
| BC151106.1 | - | 4 - 5 | 960 | 9088019 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp26c1
Genomic Map