Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200011011.mm.1 |
NM_001110239.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011011.mm.1 |
NM_001110239.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011011.mm.1 |
TC1200001589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011013.mm.1 |
NM_001110239.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011013.mm.1 |
NM_001110239.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011013.mm.1 |
NM_021330.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011013.mm.1 |
NM_021330.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011013.mm.1 |
TC1200001589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011014.mm.1 |
NM_001110239.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011014.mm.1 |
NM_001110239.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011014.mm.1 |
NM_021330.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011014.mm.1 |
NM_021330.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011014.mm.1 |
TC1200001589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011018.mm.1 |
NM_001110239.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011018.mm.1 |
NM_001110239.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011018.mm.1 |
NM_021330.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011018.mm.1 |
NM_021330.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011018.mm.1 |
TC1200001589.mm.1 |
MTA TC |