Assay Details
| Gene Symbol: Acp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110239.1 | NP_001103709.1 | 4 - 5 | 367 | 144 | |
| GenBank mRNA | AK014603.1 | - | 4 - 5 | 338 | 144 | |
| AK018329.1 | - | 4 - 5 | 346 | 144 | ||
| AK019186.1 | - | 4 - 5 | 314 | 144 | ||
| AK136144.1 | - | 4 - 5 | 344 | 144 | ||
| AK152429.1 | - | 4 - 5 | 345 | 144 | ||
| AK170284.1 | - | 4 - 5 | 323 | 144 | ||
| AK217793.1 | - | 2 - 3 | 135 | 144 | ||
| BC039744.1 | - | 4 - 5 | 368 | 3156157 | 144 | |
| BC093527.1 | - | 4 - 5 | 294 | 5137663 | 144 | |
| BC112413.1 | - | 4 - 5 | 306 | 40060984 | 144 | |
| BC145199.1 | - | 4 - 5 | 306 | 9053750 | 144 | |
| Y17343.1 | - | 4 - 5 | 287 | 144 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | Y17344.1 | - | 548 | Chr. - - [-] on Build | 144 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Acp1
Genomic Map