Assay Details
| Gene Symbol: Cdhr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006216.1 | - | 1 - 2 | 258 | 61 | |
| BG080853.2 | - | 2 - 3 | 240 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006216.1 transcripts.
Set Membership: |
|

Cdhr4
Genomic Map