Assay Details
| Gene Symbol: Cdhr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BG071036.2 | - | 2 - 3 | 87 | ||
| BG080853.2 | - | 3 - 4 | 341 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdhr4| Gene Symbol: Cdhr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BG071036.2 | - | 2 - 3 | 87 | ||
| BG080853.2 | - | 3 - 4 | 341 | 87 | ||
Set Membership: |
|