Assay Details
| Gene Symbol: Neat1 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028745.1 | - | LIT3353 VINC |
1 | 351 | 76 | |
| AK033579.1 | - | 1 | 349 | 76 | |||
| AK145725.1 | - | 1 | 349 | 76 | |||
| AK159400.1 | - | 1 | 348 | 76 | |||
| AK159773.1 | - | 1 | 351 | 76 | |||
| AK167154.1 | - | 1 | 350 | 76 | |||
| AK167255.1 | - | 1 | 349 | 76 | |||
| AK215842.1 | - | 1 | 76 | ||||
| EF177378.1 | - | 1 | 348 | 76 | |||
| GQ859163.1 | - | 348 | 76 | ||||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Neat1
Genomic Map