Assay Details
| Gene Symbol: Raxos1 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033044.1 | - | LIT3413 RaxOS |
1 - 2 | 92 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 30094 |
Gm30094 | -42.8kb | Anti-sense | Chr. 18 - 65863340 - 65894755 on NCBI Build GRCm38 | |
| Downstream | complexin 4 | Cplx4 |
A930004D23Rik, CPXIV, Gm957 |
15.3kb | Anti-sense | Chr. 18 - 65955722 - 65970178 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Raxos1
Genomic Map