Assay Details
Gene Symbol: Hoxa11os | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U20368.1 | - | LIT1566 Hoxa11as |
1 | 354 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | homeobox A11 | Hoxa11 |
Hox-1.9, Hoxa-11 |
-2.9kb | Anti-sense | Chr. 6 - 52242105 - 52245810 on NCBI Build GRCm38 |
Downstream | homeobox A13 | Hoxa13 |
Hd, Hox-1.10 |
9.1kb | Anti-sense | Chr. 6 - 52258853 - 52260880 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
