Assay Details
Gene Symbol: Ipw | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015351.1 | 168 | 86 | ||||
GenBank mRNA | AK045353.1 | - | 2 - 3 | 86 | |||
U69888.1 | - | LIT1675 IPW |
168 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
small nucleolar RNA, C/D box 116 |
Snord116 |
MBII-85, Pwcr1 |
Sense | Chr. 7 - 59675990 - 59676079 on NCBI Build GRCm38 | |
Downstream |
small nucleolar RNA, C/D box 116-like 2 |
Snord116l2 | Pwcr1l2 | 200.5kb | Sense | Chr. 7 - 59876935 - 59877024 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
