Assay Details
Gene Symbol: Ipw | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015351.1 | 605 | 154 | ||||
GenBank mRNA | U69888.1 | - | LIT1675 IPW |
605 | 154 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124856 | NR_015351.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124856 | U69888.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176358 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246363 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 690592 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814804 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51820 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88857 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 199233 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 487079 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676093 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223218 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 550011 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 941048 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969632 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191039 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531505 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 836675 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645802 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39684 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824514 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142552 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 280293 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756939 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919125 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409381 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179501 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145014 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60651 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103001 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156979 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 70641 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60411 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149162 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147389 | - | - |
Whole Mouse Genome G4122A | A_51_P411154 | AK045353.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135163 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108651 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1440557_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431229_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431229_at | - | - |
Mouse Genome 430 2.0 Array | 1431229_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1440557_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1440557_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700028479.mm.1 | NR_015351.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028479.mm.1 | NR_015351.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028479.mm.1 | TC0700004650.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028480.mm.1 | NR_015351.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028480.mm.1 | NR_015351.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028480.mm.1 | TC0700004650.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028483.mm.1 | NR_015351.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028483.mm.1 | NR_015351.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028483.mm.1 | TC0700004650.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028478.mm.1 | NR_015351.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028478.mm.1 | NR_015351.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028478.mm.1 | TC0700004650.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700028469.mm.1 | NR_015351.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028469.mm.1 | NR_015351.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700028469.mm.1 | TC0700004650.mm.1 | MTA TC |
Mouse Expression Set 430 | 1431229_PM_at | - | - |
Mouse Expression Set 430 | 1440557_at | - | - |
Mouse Expression Set 430 | 1440557_PM_at | - | - |
Mouse Expression Set 430 | 1431229_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440557_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431229_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431229_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440557_PM_at | - | - |
