Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600013709.mm.1 |
NM_001134480.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013709.mm.1 |
NM_001134480.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013709.mm.1 |
TC1600001671.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600013707.mm.1 |
NM_001134480.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013707.mm.1 |
NM_001134480.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013707.mm.1 |
TC1600001671.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600013711.mm.1 |
NM_001134480.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013711.mm.1 |
NM_001134480.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013711.mm.1 |
TC1600001671.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600013708.mm.1 |
NM_001134480.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013708.mm.1 |
NM_001134480.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013708.mm.1 |
TC1600001671.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600013710.mm.1 |
NM_001134480.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013710.mm.1 |
NM_001134480.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600013710.mm.1 |
TC1600001671.mm.1 |
MTA TC |