Assay Details
Gene Symbol: Bcl2a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007534.3 | NP_031560.2 | 1 - 2 | 508 | 112 | |
GenBank mRNA | AK138369.1 | - | 1 - 2 | 488 | 112 | |
AK152200.1 | - | 1 - 2 | 478 | 112 | ||
BC100462.1 | - | 1 - 2 | 485 | 30295442 | 112 | |
BC116241.1 | - | 1 - 2 | 508 | 40091848 | 112 |
Gene Symbol: Bcl2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009742.3 | NP_033872.1 | 1 - 2 | 552 | 112 | |
GenBank mRNA | BC028762.1 | - | 498 | 1226745 | 112 | |
CT010250.1 | - | 421 | 112 | |||
L16462.1 | - | 1 - 2 | 552 | 112 |
Gene Symbol: Bcl2a1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007536.2 | NP_031562.1 | 1 - 2 | 552 | 112 | |
GenBank mRNA | AK165576.1 | - | 1 - 2 | 304 | 112 | |
BC027536.1 | - | 1 - 2 | 465 | 1363928 | 112 | |
BC053965.1 | - | 1 - 2 | 574 | 112 | ||
BC116240.2 | - | 1 - 2 | 508 | 40091847 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
intrinsic apoptotic signaling pathway in response to DNA damage
extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of intrinsic apoptotic signaling pathway
B cell homeostasis
negative regulation of B cell apoptotic process
apoptotic process
regulation of apoptotic process
negative regulation of apoptotic process
T cell differentiation
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
T cell receptor signaling pathway
protein homodimerization activity
protein heterodimerization activity
BH domain binding
protein binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
intrinsic apoptotic signaling pathway in response to DNA damage
extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of intrinsic apoptotic signaling pathway
B cell homeostasis
negative regulation of B cell apoptotic process
apoptotic process
regulation of apoptotic process
negative regulation of apoptotic process
T cell differentiation
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
T cell receptor signaling pathway
protein homodimerization activity
protein heterodimerization activity
BH domain binding
protein binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978424 | NM_007536.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978424 | AK165576.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978424 | BC027536.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978424 | BC053965.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978424 | NM_007534.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | ERCC-00017_150 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978424 | NM_007536.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978424 | AK165576.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978424 | BC027536.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978424 | BC053965.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978424 | NM_007534.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26711182 | - | - |
Whole Mouse Genome G4122A | A_51_P494241 | NM_007534.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P494241 | AK138369.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P494241 | AK152200.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P494241 | BC100462.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P494241 | BC116241.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108560 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26711281 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711294 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P32977704 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711301 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711291 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711286 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711406 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08021398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P32977674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711182 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711282 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711273 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07530486 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07602261 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711279 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711284 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26711278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P32977749 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10587692 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10595635 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587691 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587694 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10595634 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587691 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587685 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587694 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10595635 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587684 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587685 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10595634 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587692 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587684 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10595635 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587693 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587693 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587692 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10595634 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587694 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587685 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587693 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10587684 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10587690 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10595633 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10595633 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10587690 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10587683 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10587683 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10587690 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10587683 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10595633 | - | - |
Mouse Genome 430 2.0 Array | 1419004_s_at | - | - |
Mouse Genome 430 2.0 Array | 1419004_s_at | - | - |
Mouse Genome 430 2.0 Array | 1419004_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1450812_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1437913_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1419004_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1437913_at | - | - |
Mouse Genome 430 2.0 Array | 1450812_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900023298.mm.1 | NM_007536.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900023298.mm.1 | NM_007536.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900023298.mm.1 | TC0900002806.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900023301.mm.1 | NM_007536.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900023301.mm.1 | NM_007536.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900023301.mm.1 | TC0900002806.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900008961.mm.1 | NM_007534.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900008961.mm.1 | NM_007534.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900008961.mm.1 | TC0900003331.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900023300.mm.1 | NM_007536.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900023300.mm.1 | NM_007536.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900023300.mm.1 | TC0900002806.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900008949.mm.1 | NM_009742.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900008949.mm.1 | NM_009742.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900008949.mm.1 | TC0900001142.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900023302.mm.1 | NM_007536.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900023302.mm.1 | NM_007536.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900023302.mm.1 | TC0900002806.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900028041.mm.1 | NM_007534.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900028041.mm.1 | NM_007534.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900028041.mm.1 | TC0900003331.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900008948.mm.1 | NM_009742.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900008948.mm.1 | NM_009742.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900008948.mm.1 | TC0900001142.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900008960.mm.1 | NM_007534.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900008960.mm.1 | NM_007534.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900008960.mm.1 | TC0900003331.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900023299.mm.1 | NM_007536.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900023299.mm.1 | NM_007536.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900023299.mm.1 | TC0900002806.mm.1 | MTA TC |
Mouse Expression Set 430 | 1419004_PM_s_at | - | - |
Mouse Expression Set 430 | 1437913_at | - | - |
Mouse Expression Set 430 | 1437913_at | - | - |
Mouse Expression Set 430 | 1437913_PM_at | - | - |
Mouse Expression Set 430 | 1437913_at | - | - |
Mouse Expression Set 430 | 1450812_PM_at | - | - |
Mouse Expression Set 430 | 1419004_s_at | - | - |
Mouse Expression Set 430 | 1450812_PM_at | - | - |
Mouse Expression Set 430 | 1419004_s_at | - | - |
Mouse Expression Set 430 | 1450812_at | - | - |
Mouse Expression Set 430 | 1419004_s_at | - | - |
Mouse Expression Set 430 | 1437913_PM_at | - | - |
Mouse Expression Set 430 | 1450812_at | - | - |
Mouse Expression Set 430 | 1450812_at | - | - |
Mouse Expression Set 430 | 1419004_PM_s_at | - | - |
Mouse Expression Set 430 | 1419004_PM_s_at | - | - |
Mouse Expression Set 430 | 1450812_PM_at | - | - |
Mouse Expression Set 430 | 1437913_PM_at | - | - |
Murine Genome U74v2 Set | 102914_s_at | - | - |
Murine Genome U74v2 Set | 93869_s_at | - | - |
Murine Genome U74v2 Set | 102913_PM_at | - | - |
Murine Genome U74v2 Set | 102914_PM_s_at | - | - |
Murine Genome U74v2 Set | 93869_PM_s_at | - | - |
Murine Genome U74v2 Set | 102913_at | - | - |
Murine Genome U74v2 Set | 93869_PM_s_at | - | - |
Murine Genome U74v2 Set | 102913_at | - | - |
Murine Genome U74v2 Set | 102913_at | - | - |
Murine Genome U74v2 Set | 102913_PM_at | - | - |
Murine Genome U74v2 Set | 93869_s_at | - | - |
Murine Genome U74v2 Set | 102914_PM_s_at | - | - |
Murine Genome U74v2 Set | 102914_PM_s_at | - | - |
Murine Genome U74v2 Set | 102914_s_at | - | - |
Murine Genome U74v2 Set | 102913_PM_at | - | - |
Murine Genome U74v2 Set | 102914_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1437913_at | - | - |
Mouse Genome 430A 2.0 Array | 1419004_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1419004_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1450812_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450812_at | - | - |
Mouse Genome 430A 2.0 Array | 1437913_at | - | - |
Mouse Genome 430A 2.0 Array | 1437913_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419004_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1450812_at | - | - |
Mouse Genome 430A 2.0 Array | 1437913_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419004_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1450812_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450812_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419004_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1437913_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419004_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1437913_at | - | - |
Mouse Genome 430A 2.0 Array | 1450812_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 789504 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 7000408 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2370092 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5700138 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005700138 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0007000408 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002370092 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2660263 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2743763 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213698 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1213698 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2660263 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2743763 | - | - |
mouse-6 v1.1 (Illumina) | scl0012044.1_118-S | - | - |
mouse-6 v1.1 (Illumina) | scl0012045.1_114-S | - | - |
mouse-6 v1.1 (Illumina) | scl0012047.1_125-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 7000408 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2370092 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5700138 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0012044.1_118-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0012047.1_125-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0012045.1_114-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2370092 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7000408 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5700138 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3006505 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2660263 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2660555 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3006505 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0012044.1_118-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0012045.1_114-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0012047.1_125-S | - | - |
