Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200017605.mm.1 |
NM_001097977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017605.mm.1 |
TC0200002237.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017604.mm.1 |
NM_001097977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017604.mm.1 |
TC0200002237.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017606.mm.1 |
NM_001097977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017606.mm.1 |
TC0200002237.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017608.mm.1 |
NM_001097977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017608.mm.1 |
TC0200002237.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017609.mm.1 |
NM_001097977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017609.mm.1 |
TC0200002237.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017607.mm.1 |
NM_001097977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017607.mm.1 |
TC0200002237.mm.1 |
MTA TC |