Assay Details
| Gene Symbol: Vmn2r59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105056.1 | NP_001098526.1 | 6 | 1849 | 140 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070492 | NR_028441.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070492 | AF022253.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2106700 | NM_001105056.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935985 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 942457 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 679495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341148 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369193 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700940 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576860 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670219 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538386 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306257 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 686782 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99417 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114036 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 72125 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 91290 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070492 | NR_028441.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070492 | AF022253.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2106700 | NM_001105056.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25079001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25079005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25078835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06544982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06561237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06490397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06560260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25079004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06660666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06617461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25078951 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552336 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552334 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536084 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552335 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10562641 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552341 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10536079 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552331 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10536083 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700025952.mm.1 | TC0700002916.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025952.mm.1 | NM_001105056.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025952.mm.1 | NM_001105056.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025951.mm.1 | TC0700002916.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025951.mm.1 | NM_001105056.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025951.mm.1 | NM_001105056.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025953.mm.1 | TC0700002916.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025953.mm.1 | NM_001105056.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025953.mm.1 | NM_001105056.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025954.mm.1 | TC0700002916.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025954.mm.1 | NM_001105056.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025954.mm.1 | NM_001105056.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025955.mm.1 | TC0700002916.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025955.mm.1 | NM_001105056.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025955.mm.1 | NM_001105056.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025950.mm.1 | TC0700002916.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700025950.mm.1 | NM_001105056.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700025950.mm.1 | NM_001105056.1 | Refseq |
| Murine Genome U74v2 Set | 101352_g_at | - | - |
| Murine Genome U74v2 Set | 101352_PM_g_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 876011 | - | - |

Vmn2r59
Genomic Map