Assay Details
| Gene Symbol: Hist1h2br | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC011440.1 | - | 1 - 2 | 388 | 3989862 | 88 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
histone cluster 1, H2ap |
Hist1h2ap |
ENSMUSG00000060262, Gm11276, OTTMUSG00000000448 |
-283.0 | Anti-sense | Chr. 13 - 21833024 - 21833475 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 40830 |
Gm40830 | 60.7kb | Sense | Chr. 13 - 21896116 - 21908096 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Hist1h2br
Genomic Map