Assay Details
| Gene Symbol: ChkbCpt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_004843.2 | 10 - 11 | 1780 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
carnitine palmitoyltransferase 1b, muscle |
Cpt1b |
Cpt1, Cpt1-m, Cpti, Cpti-m, M-cpti |
Sense | Chr. 15 - 89416405 - 89425862 on NCBI Build GRCm38 | |
| Downstream | mitogen-activated protein kinase 8 interacting protein 2 | Mapk8ip2 |
3230402N03Rik, AI847694, AW049958, IB2, Jip2, R75148 |
27.3kb | Anti-sense | Chr. 15 - 89453911 - 89462447 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

ChkbCpt1b
Genomic Map