Assay Details
Gene Symbol: Cdk3-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_004853.1 | 5 - 6 | 592 | 94 | ||
GenBank mRNA | AK009918.1 | - | 3 - 4 | 418 | 94 | |
BC116895.1 | - | 3 - 4 | 328 | 94 | ||
BC119034.1 | - | 3 - 4 | 328 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TEN1 telomerase capping complex subunit | Ten1 | 2310004N24Rik | -1.7kb | Sense | Chr. 11 - 116198855 - 116215318 on NCBI Build GRCm38 |
Downstream | envoplakin | Evpl | 2.3kb | Anti-sense | Chr. 11 - 116220559 - 116238147 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000001968.rn
Additional Information:
This assay is in the 5'UTR region of the AK009918.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
