Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1430573_PM_s_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1444421_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1444421_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1444421_at |
- |
- |
Mouse Genome 430 2.0 Array |
1430573_PM_s_at |
- |
- |
Mouse Genome 430 2.0 Array |
1430573_s_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1300004845.mm.1 |
NR_015349.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004845.mm.1 |
TC1300000728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004846.mm.1 |
NR_015349.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004846.mm.1 |
TC1300000728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004847.mm.1 |
NR_015349.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004847.mm.1 |
TC1300000728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004848.mm.1 |
NR_015349.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004848.mm.1 |
TC1300000728.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004849.mm.1 |
NR_015349.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004849.mm.1 |
TC1300000728.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1430573_PM_s_at |
- |
- |
Mouse Expression Set 430 |
1430573_s_at |
- |
- |
Mouse Expression Set 430 |
1444421_PM_at |
- |
- |
Mouse Expression Set 430 |
1444421_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1430573_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1430573_PM_s_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1430573_PM_s_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1430573_s_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1444421_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1444421_PM_at |
- |
- |