Assay Details
Gene Symbol: Abca16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278943.1 | NP_001265872.1 | 15 - 16 | 2487 | 147 | |
NM_001278944.1 | NP_001265873.1 | 18 - 19 | 2956 | 147 | ||
GenBank mRNA | AY243472.1 | - | 18 - 19 | 2955 | 147 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
ATP-binding cassette, sub-family A (ABC1), member 15 |
Abca15 | 4930500I12Rik | -106.8kb | Sense | Chr. 7 - 120328486 - 120407687 on NCBI Build GRCm38 |
Downstream | Y box protein 1 pseudogene | E130201H02Rik | 77.4kb | Sense | Chr. 7 - 120597625 - 120598475 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
