Assay Details
| Gene Symbol: Snora69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002900.1 | 1 | 57 | 82 | ||
| GenBank mRNA | AF357420.1 | - | 1 | 57 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sosondowah ankyrin repeat domain family member D | Sowahd |
A630014H24Rik, Ankrd58 |
-32.5kb | Anti-sense | Chr. X - 37048845 - 37050418 on NCBI Build GRCm38 |
| Downstream | UPF3 regulator of nonsense transcripts homolog B (yeast) | Upf3b |
5730594O13Rik, AI317193, AW541158, RENT3B, UPF3X |
8.6kb | Sense | Chr. X - 37091678 - 37110396 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snora69
Genomic Map