Assay Details
| Gene Symbol: Snord35a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_000003.8 | 1 | 40 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein S11 | Rps11 | -2.0kb | Sense | Chr. 7 - 45122388 - 45124389 on NCBI Build GRCm38 | |
| Downstream |
small nucleolar RNA, C/D box 34 |
Snord34 |
Rnu34, U34 |
168.0 | Sense | Chr. 7 - 45126603 - 45126668 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snord35a
Genomic Map