Assay Details
Gene Symbol: Gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027343.1 | 1 - 2 | 247 | 97 | ||
GenBank mRNA | AK168928.1 | - | 1 - 2 | 180 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glutamate decarboxylase 1 | Gad1 |
EP10, GAD25, GAD44, GAD67, Gad-1 |
-59.5kb | Sense | Chr. 2 - 70561763 - 70602014 on NCBI Build GRCm38 |
Downstream | tousled-like kinase 1 | Tlk1 | 4930545J15Rik | 39.6kb | Anti-sense | Chr. 2 - 70712407 - 70825480 on NCBI Build GRCm38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK168928.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
