Assay Details
| Gene Symbol: Hspa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027492.1 | 4 - 5 | 801 | 95 | ||
| GenBank mRNA | AK030553.1 | - | 4 - 5 | 774 | 95 | |
| BC094508.1 | - | 4 - 5 | 744 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RNA binding motif protein 11 | Rbm11 | A330018F01 | -155.8kb | Anti-sense | Chr. 16 - 75592844 - 75602825 on NCBI Build GRCm38 |
| Downstream |
SAM domain, SH3 domain and nuclear localization signals, 1 |
Samsn1 |
4930571B16Rik, 930571B16Rik, Hacs1, SLy2 |
99.2kb | Sense | Chr. 16 - 75858794 - 75909266 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Hspa13
Genomic Map