Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00000728.mm.1 |
NM_199319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000728.mm.1 |
NM_199319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000728.mm.1 |
TC0X00003387.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000751.mm.1 |
NM_001081564.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000751.mm.1 |
NM_001081564.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000751.mm.1 |
TC0X00000094.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000730.mm.1 |
NM_199319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000730.mm.1 |
NM_199319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000730.mm.1 |
TC0X00003387.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000727.mm.1 |
NM_199319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000727.mm.1 |
NM_199319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000727.mm.1 |
TC0X00003387.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000750.mm.1 |
NM_001081564.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000750.mm.1 |
NM_001081564.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000750.mm.1 |
TC0X00000094.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000725.mm.1 |
NM_199319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000725.mm.1 |
NM_199319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000725.mm.1 |
TC0X00003387.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000729.mm.1 |
NM_199319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000729.mm.1 |
NM_199319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000729.mm.1 |
TC0X00003387.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000726.mm.1 |
NM_199319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000726.mm.1 |
NM_199319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000726.mm.1 |
TC0X00003387.mm.1 |
MTA TC |