Assay Details
| Gene Symbol: Tpi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009415.2 | NP_033441.2 | 6 - 7 | 795 | 87 | |
| GenBank mRNA | AK002270.1 | - | 6 - 7 | 657 | 87 | |
| AK008034.1 | - | 2 - 3 | 155 | 87 | ||
| AK008373.1 | - | 2 - 3 | 87 | |||
| AK149768.1 | - | 6 - 7 | 659 | 87 | ||
| AK150735.1 | - | 6 - 7 | 656 | 87 | ||
| AK159741.1 | - | 6 - 7 | 659 | 87 | ||
| AK167437.1 | - | 6 - 7 | 660 | 87 | ||
| AK168060.1 | - | 6 - 7 | 656 | 87 | ||
| AK168446.1 | - | 6 - 7 | 659 | 87 | ||
| AK168756.1 | - | 6 - 7 | 658 | 87 | ||
| AK212744.1 | - | 2 - 3 | 160 | 87 | ||
| BC004024.1 | - | 6 - 7 | 643 | 87 | ||
| BC046761.1 | - | 6 - 7 | 646 | 5693696 | 87 | |
| X53333.1 | - | 6 - 7 | 657 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glucose metabolic process
gluconeogenesis
glycolytic process
pentose-phosphate shunt
multicellular organism development
metabolic process
embryo development
glyceraldehyde-3-phosphate metabolic process
catalytic activity
triose-phosphate isomerase activity
isomerase activity
ubiquitin protein ligase binding
glucose metabolic process
gluconeogenesis
glycolytic process
pentose-phosphate shunt
multicellular organism development
metabolic process
embryo development
glyceraldehyde-3-phosphate metabolic process
catalytic activity
triose-phosphate isomerase activity
isomerase activity
ubiquitin protein ligase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988424 | BC004024.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988424 | BC046761.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988424 | X53333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988424 | AK168756.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988424 | AK168446.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433824 | BC046761.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433824 | X53333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433824 | AK168756.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433824 | AK190112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433824 | AK191323.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433824 | BC046761.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433824 | X53333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433824 | AK168756.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433824 | AK190112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433824 | AK191323.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192427 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712317 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988424 | BC004024.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988424 | BC046761.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988424 | X53333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988424 | AK168756.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988424 | AK168446.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433824 | BC046761.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433824 | X53333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433824 | AK168756.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433824 | AK190112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433824 | AK191323.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433824 | BC046761.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433824 | X53333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433824 | AK168756.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433824 | AK190112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433824 | AK191323.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433824 | BC046761.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433824 | X53333.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433824 | AK168756.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433824 | AK190112.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433824 | AK191323.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433824 | BC046761.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433824 | X53333.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433824 | AK168756.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433824 | AK190112.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433824 | AK191323.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817092 | - | - |
| Whole Mouse Genome G4122A | A_52_P116102 | AK053780.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433824 | BC046761.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433824 | X53333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433824 | AK168756.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433824 | AK190112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433824 | AK191323.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433824 | BC046761.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433824 | X53333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433824 | AK168756.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433824 | AK190112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433824 | AK191323.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118993 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131592 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109467 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05259001 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05309603 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05309593 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05309591 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05258997 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05309605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05471745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24817095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24817082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05259005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24817093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05309605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24817079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24817098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24817100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24817091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05258999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24817086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05471762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24817092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05471756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05258987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24817103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05258993 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10547832 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547833 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547831 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10457730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547835 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435659_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415918_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452927_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10457729 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10547830 | - | - |
| Mouse Genome 430 2.0 Array | 1452927_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1452927_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1435659_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1435659_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1415918_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1415918_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21991_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21991_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa16190_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600024217.mm.1 | TC0600003119.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600024217.mm.1 | NM_009415.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600024217.mm.1 | NM_009415.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600024218.mm.1 | TC0600003119.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600024218.mm.1 | NM_009415.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600024218.mm.1 | NM_009415.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600024220.mm.1 | TC0600003119.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600024220.mm.1 | NM_009415.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600024220.mm.1 | NM_009415.2 | Refseq |
| Mouse Expression Set 430 | 1435659_a_at | - | - |
| Mouse Expression Set 430 | 1435659_PM_a_at | - | - |
| Mouse Expression Set 430 | 1452927_x_at | - | - |
| Mouse Expression Set 430 | 1415918_a_at | - | - |
| Mouse Expression Set 430 | 1415918_PM_a_at | - | - |
| Mouse Expression Set 430 | 1452927_PM_x_at | - | - |
| Murine Genome U74v2 Set | 99566_at | - | - |
| Murine Genome U74v2 Set | 99566_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435659_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452927_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452927_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415918_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435659_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415918_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435659_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452927_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415918_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435659_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452927_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415918_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 359347 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1500215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2100154 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1500215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001500215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002100154 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230137 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231447 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230137 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231447 | - | - |
| mouse-6 v1.1 (Illumina) | scl021991.1_200-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0021991.1_26-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1500215 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2100154 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl021991.1_200-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0021991.1_26-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1500215 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100154 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2932359 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1230137 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1231447 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231447 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2932359 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230137 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl021991.1_200-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0021991.1_26-S | - | - |

Tpi1
Genomic Map