Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100024929.mm.1 |
NM_011396.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024929.mm.1 |
NM_011396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024929.mm.1 |
TC1100002748.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100024928.mm.1 |
NM_011396.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024928.mm.1 |
NM_011396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024928.mm.1 |
TC1100002748.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100024927.mm.1 |
NM_011396.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024927.mm.1 |
NM_011396.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024927.mm.1 |
TC1100002748.mm.1 |
MTA TC |