Assay Details
| Gene Symbol: Mprip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012027.2 | NP_036157.2 | 19 - 20 | 3038 | 81 | |
| NM_201245.3 | NP_957697.1 | 19 - 20 | 3038 | 81 | ||
| GenBank mRNA | AB093269.1 | - | 7 - 8 | 4863 | 81 | |
| AK154807.1 | - | 20 - 21 | 3045 | 81 | ||
| AK155630.1 | - | 19 - 20 | 2819 | 81 | ||
| AK158296.1 | - | 19 - 20 | 3027 | 81 | ||
| AK162530.1 | - | 17 - 18 | 2384 | 81 | ||
| AK165710.1 | - | 4 - 5 | 992 | 81 | ||
| AK170849.1 | - | 19 - 20 | 3022 | 81 | ||
| AK171089.1 | - | 19 - 20 | 3027 | 81 | ||
| BC027417.1 | - | 5 - 6 | 1127 | 81 | ||
| BC039932.1 | - | 18 - 19 | 2562 | 81 | ||
| BC049803.1 | - | 19 - 20 | 2564 | 81 | ||
| BC063067.1 | - | 19 - 20 | 3001 | 5721396 | 81 | |
| U73200.1 | - | 19 - 20 | 2898 | 81 | ||
More Information
Set Membership: |
|

Mprip
Genomic Map