Assay Details
| Gene Symbol: Psmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134101.2 | NP_598862.1 | 1 - 2 | 197 | 103 | |
| GenBank mRNA | AK146482.1 | - | 1 - 2 | 146 | 103 | |
| AK150374.1 | - | 1 - 2 | 169 | 103 | ||
| AK151588.1 | - | 1 - 2 | 169 | 103 | ||
| AK152007.1 | - | 1 - 2 | 169 | 103 | ||
| AK152970.1 | - | 1 - 2 | 166 | 103 | ||
| AK159098.1 | - | 1 - 2 | 149 | 103 | ||
| AK159636.1 | - | 1 - 2 | 146 | 103 | ||
| AK159756.1 | - | 1 - 2 | 169 | 103 | ||
| AK159815.1 | - | 1 - 2 | 165 | 103 | ||
| AK166816.1 | - | 1 - 2 | 149 | 103 | ||
| AK167326.1 | - | 1 - 2 | 149 | 103 | ||
| AK167774.1 | - | 1 - 2 | 146 | 103 | ||
| AK167786.1 | - | 1 - 2 | 147 | 103 | ||
| AK167909.1 | - | 1 - 2 | 157 | 103 | ||
| AK167995.1 | - | 1 - 2 | 165 | 103 | ||
| AK168131.1 | - | 1 - 2 | 156 | 103 | ||
| AK168543.1 | - | 1 - 2 | 146 | 103 | ||
| AK169867.1 | - | 1 - 2 | 149 | 103 | ||
| BC021508.1 | - | 1 - 2 | 166 | 5347139 | 103 | |
| BC023049.1 | - | 1 - 2 | 114 | 5356268 | 103 | |
| BC024058.1 | - | 1 - 2 | 139 | 103 | ||
| BC024830.1 | - | 1 - 2 | 137 | 5358424 | 103 | |
| BC027388.1 | - | 1 - 2 | 161 | 4952935 | 103 | |
| BC027767.1 | - | 1 - 2 | 165 | 5347281 | 103 | |
| BC027822.1 | - | 1 - 2 | 148 | 5321505 | 103 | |
| BC028851.1 | - | 1 - 2 | 137 | 3660169 | 103 | |
| BC031128.1 | - | 1 - 2 | 166 | 4982313 | 103 | |
| BY307723.1 | - | 1 - 2 | 198 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023049.1 transcripts.
Set Membership: |
|

Psmd2
Genomic Map