Assay Details
Gene Symbol: 1700084F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045965.1 | 2 - 3 | 298 | 70 | ||
GenBank mRNA | AK005629.1 | - | 3 - 4 | 357 | 70 | |
AK006991.1 | - | 2 - 3 | 299 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 3772 | Gm3772 | Anti-sense | Chr. 13 - 70021959 - 70125917 on NCBI Build GRCm38 | ||
Downstream | RIKEN cDNA 4930520P13 gene | 4930520P13Rik | 206.0kb | Anti-sense | Chr. 13 - 70232822 - 70248122 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
