Assay Details
Gene Symbol: 1700084F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045965.1 | 3 - 4 | 383 | 102 | ||
GenBank mRNA | AK005629.1 | - | 4 - 5 | 442 | 102 | |
AK006991.1 | - | 3 - 4 | 384 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4933416O17 gene | 4933416O17Rik | -58.7kb | Anti-sense | Chr. 13 - 69957962 - 69962084 on NCBI Build GRCm38 | |
Downstream | predicted gene 3772 | Gm3772 | 910.0 | Anti-sense | Chr. 13 - 70021959 - 70125917 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
